missing translation for 'onlineSavingsMsg'
Learn More

Go 6983, MedChemExpress

Product Code. p-200098330
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16478477

Brand: Medchem Express HY13689

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Go 6983 is a pan-PKC inhibitor against for PKCα, PKCβ, PKCγ, PKCδ and PKCζ with IC50 of 7 nM, 7 nM, 6 nM, 10 nM and 60 nM, respectively.

Go 6983 is a pan-PKC inhibitor against for PKCα, PKCβ, PKCγ, PKCδ and PKCζ with IC50 of 7 nM, 7 nM, 6 nM, 10 nM and 60 nM, respectively.

Specifications

Chemical Name or Material Go 6983
CAS 133053-19-7
Color Red
Purity Grade Notes Research
Molecular Formula C26H26N4O3
Quantity 5 mg
Synonym Gö 6983 Goe 6983
Solubility Information DMSO : ≥ 34 mg/mL (76.83 mM)
SMILES O=C(C(C1=CN(CCCN(C)C)C2=C1C=C(OC)C=C2)=C3C4=CNC5=C4C=CC=C5)NC3=O
Molecular Weight (g/mol) 442.51
Formula Weight 442.51
Percent Purity 95.61%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.