Learn More
GNF-PF-3777, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Description
GNF-PF-3777 (8-Nitrotryptanthrin) is a potent human indoleamine 2,3-dioxygenase 2 (hIDO2) inhibitor which significantly reduces IDO2 activity with Ki of 0.97 μM.
Specifications
Specifications
| Chemical Name or Material | GNF-PF-3777 |
| Physical Form | Solid |
| CAS | 77603-42-0 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C15H7N3O4 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Synonym | 8-Nitrotryptanthrin |
| Solubility Information | DMSO : 6.4 mg/mL (21.83 mM; Need warming) |
| Show More |