missing translation for 'onlineSavingsMsg'
Learn More

GNF-PF-3777, MedChemExpress

Product Code. 16424416
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16424416

Brand: Medchem Express HY100687

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GNF-PF-3777 (8-Nitrotryptanthrin) is a potent human indoleamine 2,3-dioxygenase 2 (hIDO2) inhibitor which significantly reduces IDO2 activity with Ki of 0.97 μM.

GNF-PF-3777 (8-Nitrotryptanthrin) is a potent human indoleamine 2,3-dioxygenase 2 (hIDO2) inhibitor which significantly reduces IDO2 activity with Ki of 0.97 μM.

Specifications

Chemical Name or Material GNF-PF-3777
Physical Form Solid
CAS 77603-42-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C15H7N3O4
For Use With (Application) Neuroscience-Neuromodulation
Synonym 8-Nitrotryptanthrin
Solubility Information DMSO : 6.4 mg/mL (21.83 mM; Need warming)
SMILES O=C1N2C(C(C3=C2C=CC([N+]([O-])=O)=C3)=O)=NC4=CC=CC=C41
Molecular Weight (g/mol) 293.23
Formula Weight 293.23
Percent Purity 98.56%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.