Learn More
GNF-6231, MedChemExpress
Product Code. 16475737
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475737 5 mg 5mg
16464306 100 mg 100mg
16462998 10 mg 10mg
16459566 50 mg 50mg
16430016 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16475737 Supplier Medchem Express Supplier No. HY100408
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
MedChemExpress GNF-6231 is a potent, selective, and orally bioavailable Porcupine inhibitor that blocks Wnt signaling.
GNF-6231 is a potent, selective, and orally bioavailable Porcupine inhibitor that blocks Wnt signaling.
Specifications
Chemical Name or Material GNF-6231
CAS 1243245-18-2
Color White
Purity Grade Notes Research
Molecular Formula C24H25FN6O2
Quantity 5 mg
Solubility Information DMSO : ≥ 36 mg/mL (80.27 mM)
SMILES O=C(NC1=NC=C(N2CCN(C(C)=O)CC2)C=C1)CC3=CN=C(C4=CC(F)=NC=C4)C(C)=C3
Molecular Weight (g/mol) 448.49
Formula Weight 448.49
Percent Purity 99.81%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death