missing translation for 'onlineSavingsMsg'
Learn More

GNE-493, MedChemExpress

Product Code. p-200101661
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16454178

Brand: Medchem Express HY10811

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GNE-493 is a potent, selective, and orally available dual pan-PI3-kinase/mTOR inhibitor with IC50s of 3.4 nM, 12 nM, 16 nM, 16 nM and 32 nM for PI3Kα, PI3Kβ, PI3Kδ, PI3Kγ and mTOR.

GNE-493 is a potent, selective, and orally available dual pan-PI3-kinase/mTOR inhibitor with IC50s of 3.4 nM, 12 nM, 16 nM, 16 nM and 32 nM for PI3Kα, PI3Kβ, PI3Kδ, PI3Kγ and mTOR.

Specifications

Chemical Name or Material GNE-493
Physical Form Solid
CAS 1033735-94-2
Color Dust Gray
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C17H20N6O2S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 45 mg/mL (120.82 mM)
SMILES OC(C)(C)C1=CC2=C(S1)C(N3CCOCC3)=NC(C4=CN=C(N)N=C4)=N2
Molecular Weight (g/mol) 372.44
Formula Weight 372.44
Percent Purity 98.33%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.