Learn More
GNE-477, MedChemExpress
Product Code. 16410707
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
Product Code. 16410707
Brand: Medchem Express HY11042
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress GNE-477 is a potent and efficacious dual PI3K (IC50=4 nM)/mTOR(Ki=21 nM) inhibitor.
GNE-477 is a potent and efficacious dual PI3K (IC50=4 nM)/mTOR(Ki=21 nM) inhibitor.
Specifications
Chemical Name or Material GNE-477
CAS 1032754-81-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H28N8O3S2
Quantity 50 mg
Solubility Information DMSO : 16.67 mg/mL (33.03 mM; Need ultrasonic)
SMILES NC1=NC=C(C2=NC(N3CCOCC3)=C4C(C(C)=C(CN5CCN(S(=O)(C)=O)CC5)S4)=N2)C=N1
Molecular Weight (g/mol) 504.63
Formula Weight 504.63
Percent Purity 98.7%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease