missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNB2 Antibody (G4) [Janelia Fluor« 549], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
GNB2 Monoclonal antibody specifically detects GNB2 in Human,Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | GNB2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | G4 |
| Conjugate | Janelia Fluor 549 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | beta-2 subunit, guanine nucleotide binding protein (G protein), beta polypeptide 2, guanine nucleotide-binding protein G(I)/G(S)/G(T) beta subunit 2, guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-2, signal-transducing guanine nucleotide-binding regulatory protein beta subunit |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide derived from mouse GNB2 (RKACGDSTLTQITAGLD) |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?