missing translation for 'onlineSavingsMsg'
Learn More

GlyRS-IN-1, MedChemExpress

Product Code. 16444248
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16444248 10 mg 10mg
16466967 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16444248 Supplier Medchem Express Supplier No. HY108940
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress GlyRS-IN-1 is a glycyl-tRNA synthase (GlyRS) inhibitor extracted from patent WO 2017066459 A1. GlyRS-IN-1 can also inhibit the growth of bacteria.

GlyRS-IN-1 is a glycyl-tRNA synthase (GlyRS) inhibitor extracted from patent WO 2017066459 A1. GlyRS-IN-1 can also inhibit the growth of bacteria.

Specifications

Chemical Name or Material GlyRS-IN-1
CAS 112921-11-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C12H17N7O7S
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (247.91 mM; Need ultrasonic)
SMILES O[C@@H]1[C@@H](COS(=O)(NC(CN)=O)=O)O[C@@H](N2C(N=CN=C3N)=C3N=C2)[C@@H]1O
Molecular Weight (g/mol) 403.37
Formula Weight 403.37
Percent Purity 98.14%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.