Learn More
Glyphosate-C3-d2, TRC
Specifications
Specifications
| Chemical Name or Material | Glyphosate-C3-d2 |
| CAS | 2733532-11-9 |
| Molecular Formula | C3 D2 H6 N O5 P |
| Quantity | 2.5 mg |
| Synonym | N-(Phosphonomethyl)glycine-C3-d2, (Carboxymethylamino)methylphosphonic Acid-C3-d2, Phosphonomethylglycine-C3-d2, Accord-C3-d2, Glyphomax-C3-d2, Rebel Garden-C3-d2, Roundup Max-C3-d2, Silglif-C3-d2, Glyphosate-d2, Glyphosate-D2 (phosphonomethyl-D2), N-(Phosphonomethyl-d2)glycine, N-Phosphonomethyl-d2-glycine |
| InChI Formula | InChI=1S/C3H8NO5P/c5-3(6)1-4-2-10(7,8)9/h4H,1-2H2,(H,5,6)(H2,7,8,9)/i2D2 |
| SMILES | [2H]C([2H])(NCC(=O)O)P(=O)(O)O |
| IUPAC Name | 2-[[dideuterio(phosphono)methyl]amino]acetic acid |
| Molecular Weight (g/mol) | 171.09 |
| Formula Weight | 171.0266 |