Learn More
Glyphosate-C2-d2, TRC
Specifications
Specifications
| Chemical Name or Material | Glyphosate-C2-d2 |
| CAS | 2734001-36-4 |
| Molecular Formula | C3H6D2NO5P |
| Quantity | 1 mg |
| Synonym | N-(Phosphonomethyl)glycine-C2-d2, (Carboxymethylamino)methylphosphonic Acid-C2-d2, Phosphonomethylglycine-C2-d2, Accord-C2-d2, Glyphomax-C2-d2, Rebel Garden-C2-d2, Roundup Max-C2-d2, Silglif-C2-d2 |
| InChI Formula | InChI=1S/C3H8NO5P/c5-3(6)1-4-2-10(7,8)9/h4H,1-2H2,(H,5,6)(H2,7,8,9)/i1D2 |
| SMILES | OC(C([2H])([2H])NCP(O)(O)=O)=O |
| IUPAC Name | (phosphonomethyl)glycine-2,2-d2 |
| Molecular Weight (g/mol) | 171.09 |
| Formula Weight | 171.03 |