Learn More
Glyphosate-13C,15N, TRC
Specifications
Specifications
| Chemical Name or Material | Glyphosate 2-13C, 15N |
| CAS | 285978-24-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | 13C C2 H8 15N O5 P |
| Quantity | 5 mg |
| Synonym | Glyphosate 13C 15N (2-13C) |
| InChI Formula | InChI=1S/C3H8NO5P/c5-3(6)1-4-2-10(7,8)9/h4H,1-2H2,(H,5,6)(H2,7,8,9)/i1+1,4+1 |
| SMILES | OC(=O)[13CH2][15NH]CP(=O)(O)O |
| IUPAC Name | 2-(phosphonomethylamino)acetic acid |
| Molecular Weight (g/mol) | 171.06 |
| Show More |