missing translation for 'onlineSavingsMsg'
Learn More

Glycolic Acid, TRC

Product Code. 17942935
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17942935 1 g 1g
30134325 100 g 100g
30136249 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17942935 Supplier Toronto Research Chemicals Supplier No. TRCG6456001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-14-1
Formula Weight 75.04
IUPAC Name Glycolate
Molecular Formula C2H3O3-
Molecular Weight (g/mol) 75.04
Percent Purity >95%
SMILES OCC(=O)O
Synonym Acetic acid, 2-hydroxy- (ACI), 2-Hydroxyacetic acid (ACI), Acetic acid, hydroxy- (9CI), Glycolic acid (7CI, 8CI), 2-Hydroxyethanoic acid, Glycocide, GlyPure, GlyPure 70, GlyPure 99, Hydroxyacetic acid, Hydroxyethanoic acid, NSC 166, YST-GCS, α-Hydroxyacetic acid
Chemical Name or Material Glycolate
Grade GC

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.