Learn More
Glycine-2,2-d2-N-FMOC, CDN
Specifications
Specifications
| Chemical Name or Material | N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]glycine-2,2-D2 |
| CAS | 284665-11-8 |
| Molecular Formula | C17 D2 H13 N O4 |
| Quantity | 0.25 g |
| Synonym | Glycine-2,2-d2, N-[(9 H-fluoren-9-ylmethoxy)carbonyl]- (9 CI), 2,2-Dideuterio-2-(9 H-fluoren-9-ylmethoxycarbonylamino)acetic acid, N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]glycine-2,2-D2, Fmoc-[2H2]Gly-OH, N-Fmoc-glycine-d2, Glycine-2,2-d2-N-FMOC |
| InChI Formula | InChI=1 S/C17H15NO4/c19-16(20)9-18-17(21)22-10-15-13-7-3-1-5-11(13)12-6-2-4-8-14(12)15/h1-8,15 H,9-10H2,(H,18,21)(H,19,20)/i9D2 |
| SMILES | [2 H]C([2 H])(NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O |
| IUPAC Name | 2,2-dideuterio-2-(9 H-fluoren-9-ylmethoxycarbonylamino)acetic acid |
| Molecular Weight (g/mol) | 299.3176 |
| Formula Weight | 299.1127 g/mol |
| Show More |