Learn More
Glycine-13C2,15N p-Toluenesulfonate, TRC
Specifications
Specifications
| Chemical Name or Material | Glycine-13C2,15N p-Toluenesulfonate |
| CAS | 2727323-57-9 |
| Molecular Formula | 13C2H515NO2 . C7H8O3S |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C7H8O3S.C2H5NO2/c1-6-2-4-7(5-3-6)11(8,9)10;3-1-2(4)5/h2-5H,1H3,(H,8,9,10);1,3H2,(H,4,5)/i;1+1,2+1,3+1 |
| SMILES | O[13C]([13CH2][15NH2])=O.CC1=CC=C(C=C1)S(=O)(O)=O |
| Molecular Weight (g/mol) | 250.25 |
| Formula Weight | 250.06 |