Learn More
Glyceryl Triacetate-d9, CDN
Specifications
Specifications
| Chemical Name or Material | Triacetin-d9 |
| CAS | 2733570-86-8 |
| Molecular Formula | C9 D9 H5 O6 |
| Quantity | 1 g |
| Synonym | 1,2,3-Propanetriol tri(acetate-d3), 1,2,3-Propanetriol triacetate-d9, Triacetin-d9, Glycerol tri(acetate-d3), Tri(acetyl-d3)glycerol, Triacetin-D9 (triacetyl-D9) |
| InChI Formula | InChI=1 S/C9H14O6/c1-6(10)13-4-9(15-8(3)12)5-14-7(2)11/h9H,4-5H2,1-3H3/i1D3,2D3,3D3 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)OCC(COC(=O)C([2 H])([2 H])[2 H])OC(=O)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3-bis[(2,2,2-trideuterioacetyl)oxy]propyl 2,2,2-trideuterioacetate |
| Molecular Weight (g/mol) | 227.2593 |
| Formula Weight | 227.1355 g/mol |
| Show More |