Learn More
Glyceryl Tri(hexadecanoate-16,16,16-d3), CDN
Specifications
Specifications
| Chemical Name or Material | Tripalmitin-D9 (tri(hexadecanoyl-16,16,16-D3)) |
| CAS | 285979-78-4 |
| Molecular Formula | C51 D9 H89 O6 |
| Quantity | 0.05 g |
| Synonym | Hexadecanoic-16,16,16-d3 acid, 1,2,3-propanetriyl ester (9 CI), Hexadecanoic-16,16,16-d3 acid 1,2,3-propanetriyl ester, Glyceryl Tri(hexadecanoate-16,16,16-d3), Glycerol trihexadecanoate-d9, Glyceryl tripalmitate-d9, Trihexadecanoin-d9, Tripalmitin-d9, Tripalmitin-D9 (tri(hexadecanoyl-16,16,16-D3)) |
| InChI Formula | InChI=1 S/C51H98O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-49(52)55-46-48(57-51(54)45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)47-56-50(53)44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h48H,4-47H2,1-3H3/i1D3,2D3,3D3 |
| SMILES | [2 H]C([2 H])([2 H])CCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC([2 H])([2 H])[2 H])OC(=O)CCCCCCCCCCCCCCC([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3-bis(16,16,16-trideuteriohexadecanoyloxy)propyl 16,16,16-trideuteriohexadecanoate |
| Molecular Weight (g/mol) | 816.3757 |
| Formula Weight | 815.7928 g/mol |
| Show More |