missing translation for 'onlineSavingsMsg'
Learn More

β-Glycerophosphate disodium salt pentahydrate, MedChemExpress

Product Code. p-200100215
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422429 1 g 1g
16401198 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16422429 Supplier Medchem Express Supplier No. HYD0886

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress β-Glycerophosphate disodium salt pentahydrate is a phosphatase inhibitor.

β-Glycerophosphate disodium salt pentahydrate is a phosphatase inhibitor.

Specifications

Chemical Name or Material β-Glycerophosphate disodium salt pentahydrate
CAS 13408-09-8
Color White
Purity Grade Notes Research
Molecular Formula C3H17Na2O11P
Quantity 1 g
Solubility Information H2O : 250 mg/mL (816.70 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES OCC(OP(O[Na])(O[Na])=O)CO.[5H2O]
Molecular Weight (g/mol) 306.11
Formula Weight 306.11
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.