Learn More
Invitrogen™ Glycerol kinase Polyclonal Antibody

Description
Positive test controls include: DU145, U-87MG. The target is usually found in the following locations: Cytoplasm, Cytoplasmic side, Mitochondrion outer membrane, Peripheral membrane protein. Immunogen sequence: GIPLSHLQVD GGMTSNKILM QLQADILYIP VVKPSMPETT ALGAAMAAGA AEGVGVWSLE PEDLSAVTME RFEPQINAEE SEIRYSTWKK AVMKSMGWVT TQSPESGDPS IFCSLPLGFF IVSSMVMLIG ARYISGIP.
Specifications
Specifications
| Antigen | Glycerol kinase |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.04 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | GK |
| Gene Accession No. | P32189, Q63060, Q64516 |
| Gene Alias | ASTP; ATP:glycerol 3-phosphotransferase; ATP-stimulated glucocorticoid-receptor translocation promoter; ATP-stimulated glucocorticoid-receptor translocaton promoter; D930012N15Rik; EC 2.7.1.30; GK; GK 3; GK1; GK3P; GKD; GKl; GKP3; GKTB; glycerokinase; Glycerokinase 3; glycerol kinase; Glycerolkinase; Gyk |
| Gene Symbols | GK |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.