Learn More
Glutaminase C-IN-1, MedChemExpress
Product Code. 16465288
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465288 10 mg 10mg
16431497 50 mg 50mg
16428007 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16465288 Supplier Medchem Express Supplier No. HY12682
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Glutaminase C-IN-1 (Compound 968) is an allosteric inhibitor of Glutaminase C that inhibits cancer cell growth without affecting their normal cellular counterparts.
Glutaminase C-IN-1 (Compound 968) is an allosteric inhibitor of Glutaminase C that inhibits cancer cell growth without affecting their normal cellular counterparts.
Specifications
Chemical Name or Material Glutaminase C-IN-1
CAS 311795-38-7
Color White
Purity Grade Notes Research
Molecular Formula C27H27BrN2O
Quantity 10 mg
Synonym Compound 968
Solubility Information DMSO : 25 mg/mL (52.59 mM; Need ultrasonic)
SMILES O=C1C(C(C2=CC=C(N(C)C)C(Br)=C2)NC3=C4C5=CC=CC=C5C=C3)=C4CC(C)(C)C1
Molecular Weight (g/mol) 475.42
Formula Weight 475.42
Percent Purity 99.59%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death