missing translation for 'onlineSavingsMsg'
Learn More

GluR2 Antibody (1059613) [PE], Novus Biologicals™

Product Code. 30676818 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30676818 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30676818 Supplier Novus Biologicals Supplier No. FAB11333P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GluR2 Monoclonal antibody specifically detects GluR2 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR2
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 1059613
Conjugate PE
Formulation PBS
Gene Alias AMPA 2, glutamate receptor, ionotropic, AMPA 2
Host Species Mouse
Immunogen Human GluR2/GRIA2 containing peptide, Accession # P42262
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Tumor Suppressors, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2891
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.