missing translation for 'onlineSavingsMsg'
Learn More

GluR1 Antibody (S355-1), DyLight 350, Novus Biologicals™

Product Code. 18441057 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18441057 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18441057 Supplier Novus Biologicals Supplier No. NBP222399UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GluR1 Monoclonal antibody specifically detects GluR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR1
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone S355-1
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias AMPA 1, AMPA-selective glutamate receptor 1, GluA1, GLUH1, GluR-1, GLUR1gluR-A, GluR-A, GLURA, GluR-K1, glutamate receptor 1, Glutamate receptor ionotropic, AMPA 1, glutamate receptor, ionotropic, AMPA 1, HBGR1, MGC133252
Host Species Mouse
Immunogen Fusion protein amino acids 1-389 (extracellular N-terminus) of rat GluA1/GluR1
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2890
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.