missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucosylceramidase/GBA Antibody (OTI4G4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45829
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
Glucosylceramidase/GBA Monoclonal specifically detects Glucosylceramidase/GBA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Glucosylceramidase/GBA | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
| Acid beta-glucosidase, alglucerase, Beta-glucocerebrosidase, D-glucosyl-N-acylsphingosine glucohydrolase, EC 3.2.1.45, GBA1, GC, GCB, GLUC, glucosidase, beta, acid, glucosidase, beta; acid (includes glucosylceramidase), glucosylceramidase, imiglucerase, lysosomal glucocerebrosidase | |
| Mouse | |
| 55.5 kDa | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 2629 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Immunohistochemistry (Paraffin), Immunohistochemistry | |
| OTI4G4 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| GBA | |
| Human recombinant protein fragment corresponding to amino acids 40-315 of human GBA (NP_000148) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction