missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucosylceramidase/GBA Antibody (OTI4G4), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70803AF700
This item is not returnable.
View return policy
Description
Glucosylceramidase/GBA Monoclonal specifically detects Glucosylceramidase/GBA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Glucosylceramidase/GBA | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GBA | |
| Recombinant protein fragment corresponding to amino acids 40-315 of human Glucosylceramidase/GBA (NP_000148) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI4G4 | |
| Western Blot, Immunohistochemistry | |
| Acid beta-glucosidase, alglucerase, Beta-glucocerebrosidase, D-glucosyl-N-acylsphingosine glucohydrolase, EC 3.2.1.45, GBA1, GC, GCB, GLUC, glucosidase, beta, acid, glucosidase, beta; acid (includes glucosylceramidase), glucosylceramidase, imiglucerase, lysosomal glucocerebrosidase | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 2629.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction