missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucosylceramidase/GBA Antibody (2E2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00002629-M01
This item is not returnable.
View return policy
Description
Glucosylceramidase/GBA Monoclonal antibody specifically detects Glucosylceramidase/GBA in Human, Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| Glucosylceramidase/GBA | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Acid beta-glucosidase, alglucerase, Beta-glucocerebrosidase, D-glucosyl-N-acylsphingosine glucohydrolase, EC 3.2.1.45, GBA1, GC, GCB, GLUC, glucosidase, beta, acid, glucosidase, beta, acid (includes glucosylceramidase), glucosylceramidase, imiglucerase, lysosomal glucocerebrosidase | |
| GBA (NP_000148, 146 a.a. ~ 235 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SYFSEEGIGYNIIRVPMASCDFSIRTYTYADTPDDFQLHNFSLPEEDTKLKIPLIHRALQLAQRPVSLLASPWTSPTWLKTNGAVNGKGS | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 2629 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 2E2 | |
| Western Blot 1:500, ELISA, Immunohistochemistry 3ug/mL, Immunocytochemistry/ Immunofluorescence 10ug/mL, Immunoprecipitation, Immunohistochemistry-Paraffin 3ug/mL | |
| NP_000148 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human, Plant | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction