missing translation for 'onlineSavingsMsg'
Learn More

Glucoraphanin, MedChemExpress

Product Code. 16431738
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16431738 5 mg 5mg
16499298 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16431738 Supplier Medchem Express Supplier No. HYN4068
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Glucoraphanin, a natural glucosinolate found in cruciferous vegetable, is a stable precursor of the Nrf2 inducer sulforaphane, which possesses antioxidant, anti-inflammatory, and anti-carcinogenic effects.

Glucoraphanin, a natural glucosinolate found in cruciferous vegetable, is a stable precursor of the Nrf2 inducer sulforaphane, which possesses antioxidant, anti-inflammatory, and anti-carcinogenic effects.

Specifications

Chemical Name or Material Glucoraphanin
CAS 21414-41-5
Color White
Purity Grade Notes Research
Molecular Formula C12H23NO10S3
Quantity 5 mg
Solubility Information H2O : ≥ 76.5 mg/mL (174.85 mM)
SMILES CS(CCCC/C(S[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)CO)=N\OS(=O)(O)=O)=O
Molecular Weight (g/mol) 437.51
Formula Weight 437.51
Percent Purity 98.21%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.