Learn More
Glucocorticoid receptor Mouse anti-Human, Unconjugated, Clone: 1D9A9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66904-1-IG-150UL
 View more versions of this product
Product Code. 16859185
  • 410.00 € / 150µL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
The Glucocorticoid receptor antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Glucocorticoid receptor. This antibody recognizes human, mouse, rat antigen. The Glucocorticoid receptor antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
Glucocorticoid receptor
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1D9A9
Unconjugated
150 μL
2908