Learn More
GLRX3 Monoclonal Antibody (OTI4C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811096
Beskrivning
This gene encodes a member of the glutaredoxin family. Glutaredoxins are oxidoreductase enzymes that reduce a variety of substrates using glutathione as a cofactor. The encoded protein binds to and modulates the function of protein kinase C theta. The encoded protein may also inhibit apoptosis and play a role in cellular growth, and the expression of this gene may be a marker for cancer. Pseudogenes of this gene are located on the short arm of chromosomes 6 and 9. Alternatively spliced transcript variants have been observed for this gene.Specifikationer
| GLRX3 | |
| Monoclonal | |
| Unconjugated | |
| GLRX3 | |
| GLRX4, GRX3, GRX4, PICOT, TXNL2, TXNL3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10539 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI4C8 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O76003 | |
| GLRX3 | |
| Full length human recombit protein of human GLRX3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.