Learn More
GLRA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13831-1-AP-150UL
This item is not returnable.
View return policy
Description
Glycine is an inhibitory neurotransmitter in the central nervous system. The glycine receptor (GlyR) is a member of the nicotinic acetylcholine receptor of ligand-gated ion channels. This receptor has important roles in a variety of physiological processes, especially in mediating inhibitory neurotransmission in the spinal cord and brain stem. GlyR is a pentameric receptor comprised of alpha and beta subunits. Four alpha-subunits (GLRA1, GLRA2, GLRA3, GLRA4) and one beta-subunit (GLRB) of GlyR have been identified.Specifications
| GLRA2 | |
| Polyclonal | |
| Unconjugated | |
| GLRA2 | |
| GLR, GLRA2, glycine receptor, alpha 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 237213, 24397, 2742 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P22771, P23416, Q7TNC8 | |
| GLRA2 | |
| GLRA2 Fusion Protein Ag4743 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |