Learn More
Tocris Bioscience™ GLPG 0187
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
GLPG 0187 is a potent αv integrin inhibitor (IC50 values are 1.2-3.7 nM). Inhibits osteoclastic bone resorption and angiogenesis in vitro and in vivo. Suppresses epithelial-mesenchymal transition (EMT) and migration of PC-3M-Pro4/luc cells, without inhibiting cell growth. Induces detachment and necrosis of GL-261 and SMA-560 murine glioma cells.
Specifications
Specifications
| Chemical Name or Material | 3-[[2,5-Dimethyl-6-[4-(5,6,7,8-tetrahydro-1,8-naphthyridin-2-yl)-1-piperidinyl]-4-pyrimidinyl]amino]-N-[(4-methoxyphenyl)sulfonyl]-L-alanine |
| CAS | 1320346-97-1 |
| Quantity | 10 mg |
| Target | Integrin Receptor Inhibitors |
| Molecular Formula | C29H37N7O5S |
| Purity | 0.97 |