Learn More
Hirschmann™ Class A Volumetric Flasks with Rim
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
10 L
Unit Size:
Each
Pack of 2
Description
- With blue graduation
- Acc. to DIN EN ISO 1042
- Corrosion-free and resistant to thermal shock
- Conformity certified, with dated batch identification
Specifications
Specifications
| Certifications/Compliance | DIN EN ISO 1042 |
| Capacity (Metric) | 200 mL |
| Material | Borosilicate Glass, Duran Glass |
| Height (Metric) | 210 mm |
| Limit of Error | ±0.15 mL |
| Tolerance | ±0.15 mL |
| Graduated | Yes |
| Class | A |