Learn More
Biochrom Glass Test Tubes
Description
- Glass test tubes
- 12 mm dia. x 100 mm L
- For use with Ultrospec 7000/8000/9000
- Requires Tube Holder 80-7100-11
- For use with Novaspec III/Plus
- Requires Tet Tube Adapter 80-2117-47
Specifications
Specifications
| Type | Test Tube |
| Material | Glass |
| Length (Metric) | 100 mm |
| Quantity | 10 Pk. |
| Diameter (Metric) | 12 mm |