Learn More
Horst GLASS SILK CORD 450°C
Brand:  Horst 100402
GLASS SILK CORD 450°C
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10007101
  • ONLINE EXCLUSIVE PRICE
    2.89 €
    2.50 € / 1m
    Save 0.39 €
    13% Off
Only null left
Estimated Shipment: 13-02-2026