missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Assistent™ Wide Neck Screw Cap Bottles

Product Code. 10043771
Click to view available options
Capacity (Metric):
25 mL
100 mL
1000 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10043771

Brand: Karl Hecht™ 41324006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ideal for the storage of dry powders in the laboratory. Karl Hecht™ Assistent™ Wide Neck Screw Cap Bottles allow you to access their contents with ease.

  • With thread and screw cap
  • Clear glass
  • Screw caps are heat resistant up to 110°C

Specifications

Capacity (Metric) 25 mL
Color Clear
Diameter (Metric) Inner Neck 28 mm
Includes Screw Cap
Material Glass
Volume (Metric) Dispense 25mL
Mouth Wide
Product Type Reagent Bottle
Closure Type Screw Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.