Learn More
Leica Microsystems Glass Plate
Click to view available options
Color:
Clear
Unit Size:
Each
Description
Offering incredible versatility, the S Series microscopes can be specified on various stand types with a wide range of lighting options. They are configured as fully working units with all the required elements included.Maximum 36.5mm field of view - reduced sample handling and inspection times Versions with 38° or 60° viewing angle - promotes proper posture and improved comfort Ergonomic objectives for optimised viewing height Transmitted light base with adjustable reflector - flexible and cost effective illumination LED lighting for minimum maintenance and daylight color quality Large working distance (110mm) allows ample space for sample manipulation Built to last with Leica quality Compact design for crowded labs With the S6D camera port version, images can be taken and recorded for future reference All S-series systems feature:
- One fixed, one adjustable eyepiece, both 10x magnification
- 38° viewing angle
- Total magnification of between 6.3x and 40x, continuous zoom
- Power cable and dust cover
- Lighting as per description For routine stereomicroscope work in laboratories.
Order Info
Color
Specifications
Specifications
| Type | Glass Plate |
| Color | Clear |
| Material | Glass |
| Quantity | 1 Ea. |