Learn More
Karl Hecht™ Petri Dish, Glass
Click to view available options
Diameter (Metric):
4 cm
8 cm
12 cm
15 cm
20 cm
Height (Metric):
1.2 cm
1.5 cm
2 cm
2.5 cm
5 cm
Unit Size:
Each
Description
- Glass construction.
- Lidded.
- Available in a variety of sizes.
Specifications
Specifications
| Material | Glass |
| Shape | Round |
| Type | Petri Dish |
| Height (Metric) | 2 cm |
| Diameter (Metric) | 12 cm |