Learn More
Trajan Scientific and Medical™ Glass Lined Stainless-Steel GC Tubing
Exhibits resistance to the harshest of conditions and to strong acids and bases
Brand:  Trajan Scientific and Medical™ 082712
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15218993
  • 42.70 € / Each
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
  • Ideal for HPLC columns, particularly for biological matrices
  • Temperature stable up to 500°C
Specifications
Specifications
HPLC Columns, Biological Matrices
30 cm
Stable up to 500°C
0.4 mm
Borosilicate Glass (Lining)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.