missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Assistent™ Glass Instrument Trays

Sterilizable in hot air up to +180°C. Karl Hecht™ Assistent™ Glass Instrument Trays are excellent for cleaning, sterilizing and storing laboratory instruments.

Brand:  Karl Hecht™ 42910004

Additional Details : Weight : 0.11100kg

 View more versions of this product

Product Code. 10528701

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Autoclavable
  • Available in a range of sizes
  • Available with or without stainless steel cover
Specifications

Specifications

220 x 150 x 55 mm
220 mm
Tray
With cover
55 mm
Glass, Stainless Steel
150 mm
Resistant to hot-air sterilization up to +180°C / +356°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions