Learn More
Brady™ Pneumatic Quick-Disconnect Lockout
Isolates pneumatic energy without the expense and inconvenience of installing in-line lockout valves.
Brand: Brady™ 065645
Additional Details : Weight : 0.15880kg
Description
- Device is fitted to the male fitting, isolating equipment from all compressed air sources
- Accommodates the majority of 6mm, 9mm and 12mm male fittings
Specifications
Pneumatic Shutoff | |
Glass-filled Nylon | |
6 Pk. |
Red | |
Pneumatic Quick-Disconnect Lockout |