Learn More
Gerresheimer™ Threaded Brown Wide Mouth Bottles without Closure
Product Code. 15222409
Change view
Click to view available options
Capacity (Metric):
30 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 12
Pack of 126
Pack of 20
Pack of 42
Pack of 63
Pack of 85
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15222409 250 mL Pack of 42
15272419 30 mL Pack of 126
15292419 50 mL Pack of 85
15202429 100 mL Pack of 63
15201559 500 mL Pack of 20
15252429 1000 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15222409 Supplier Gerresheimer™ Supplier No. 4446128003
Add to basket
Edge
Add to basket
Stock the laboratory with Gerresheimer™ Threaded Brown Wide Mouth Bottles without Closure. The round, glass bottles are available in various capacities.
  • Round
  • Wide Mouth
  • Brown Glass
  • Without closure
Specifications
Capacity (Metric) 250 mL
Color Brown
Material Glass
No. per Pack 42 Pack
Mouth Wide
Shape Round
Product Type Threaded Bottle