missing translation for 'onlineSavingsMsg'
Learn More

GIRK2 Antibody (KCNJ6/7558), Novus Biologicals™

Product Code. 18647014 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18647014 100 μg 100µg
18611725 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18647014 Supplier Novus Biologicals Supplier No. NBP323852100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GIRK2 Monoclonal antibody specifically detects GIRK2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIRK2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KCNJ6/7558
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Formulation 10 mM PBS with 0.05% BSA
Gene Alias BIR1Kir3.2, G protein-activated inward rectifier potassium channel 2, GIRK-2, GIRK2KATP-2, hiGIRK2, Inward rectifier K(+) channel Kir3.2, inward rectifier potassium channel KIR3.2, KATP2human G protein coupled inward rectifier potassium channel 2 (hiGIRK2)10KCNJ7KIR3.2, MGC126596, Potassium channel, inwardly rectifying subfamily J member 6, potassium inwardly-rectifying channel, subfamily J, member 6
Host Species Mouse
Immunogen Recombinant fragment of human GIRK2 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3763
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.