missing translation for 'onlineSavingsMsg'
Learn More

Girdin Polyclonal Antibody
GREENER_CHOICE

Product Code. 12879012
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12879012 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12879012 Supplier Invitrogen Supplier No. PA520869

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Girdin Polyclonal Antibody

Girdin Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen Girdin
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CCDC88A
Gene Accession No. Q3V6T2, Q5SNZ0
Gene Alias A430106J12Rik; AI848406; Akt phosphorylation enhancer; AKT-phosphorylation enhancer; Ape; C130096N06Rik; C330012F17Rik; Ccdc88a; coiled coil domain containing 88A; coiled-coil domain containing 88A; coiled-coil domain-containing protein 88A; D130005J21Rik; G alpha-interacting vesicle-associated protein; Galpha-interacting vesicle-associated protein; girders of actin filament; girders of actin filaments; girdin; GIV; Grdn; HkRP1; Hook-related protein 1; KIAA1212; RGD1306694
Gene Symbols CCDC88A
Host Species Rabbit
Immunogen A 16 amino acid peptide from near the carboxy terminus human Girdin.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108686, 305605, 55704
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.