Learn More
Girard's Reagent T, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | Girard's Reagent T |
| CAS | 123-46-6 |
| Molecular Formula | C5H14ClN3O |
| Quantity | 100 g |
| Synonym | 2-Hydrazinyl-N,N,N-trimethyl-2-oxo-ethanaminium Chloride, (Carboxymethyl)trimethylammonium Chloride Hydrazide, (Carboxymethyl)trimethyl-ammonium Chloride Hydrazide, 2-Hydrazino-N,N,N-trimethyl-2-oxo-ethanaminium Chloride, (2-Hydrazino-2-oxoethyl)trimethylammonium Chloride, (Carbazoylmethyl)trimethylammonium Chloride, Betaine Hydrazide Chloride, Betaine Hydrazide Hydrochloride, Girard-T Reagent, Girard's T Reagent, Girard's T Reagents, Trimethylacetylhydrazide Ammonium Chloride, Trimethylaminoacetohydrazide Chloride, Trimethylammonioacetohydrazide Chloride, Trimethylammonium Acetyl Hydrazide Chloride, Trimethylammonium Chloride Acetylhydrazide |
| InChI Formula | InChI=1S/C5H13N3O.ClH/c1-8(2,3)4-5(9)7-6;/h4,6H2,1-3H3;1H |
| SMILES | [Cl-].C[N+](C)(C)CC(=O)NN |
| IUPAC Name | (2-hydrazinyl-2-oxoethyl)-trimethylazanium;chloride |
| Molecular Weight (g/mol) | 167.64 |
| Formula Weight | 167.0825 |