missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ginsenoside Ro, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Ginsenoside Ro (Polysciasaponin P3
Specifications
Specifications
| Chemical Name or Material | Ginsenoside Ro |
| CAS | 34367-04-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C48H76O19 |
| Quantity | 10 mg |
| Synonym | Polysciasaponin P3 Chikusetsusaponin 5 Chikusetsusaponin V |
| Solubility Information | DMSO : 100 mg/mL (104.48 mM; Need ultrasonic) |
| SMILES | O=C([C@]1(CCC(C)(C)C2)[C@]2([H])C3=CC[C@@]4([H])[C@@](C)(CC[C@]5([H])[C@@]4(CC[C@H](O[C@@](O[C@H](C(O)=O)[C@@H](O)[C@@H]6O)([H])[C@@H]6O[C@]([C@@H]([C@@H](O)[C@@H]7O)O)([H])O[C@@H]7CO)C5(C)C)C)[C@]3(C)CC1)O[C@@H]([C@@H]([C@@H](O)[C@@H]8O)O)O[C@@H]8CO |
| Molecular Weight (g/mol) | 957.11 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction