Learn More
Ginsenoside Ro, MedChemExpress
Description
Ginsenoside Ro (Polysciasaponin P3
Specifications
Specifications
| Chemical Name or Material | Ginsenoside Ro |
| CAS | 34367-04-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C48H76O19 |
| Quantity | 10 mg |
| Synonym | Polysciasaponin P3 Chikusetsusaponin 5 Chikusetsusaponin V |
| Solubility Information | DMSO : 100 mg/mL (104.48 mM; Need ultrasonic) |
| SMILES | O=C([C@]1(CCC(C)(C)C2)[C@]2([H])C3=CC[C@@]4([H])[C@@](C)(CC[C@]5([H])[C@@]4(CC[C@H](O[C@@](O[C@H](C(O)=O)[C@@H](O)[C@@H]6O)([H])[C@@H]6O[C@]([C@@H]([C@@H](O)[C@@H]7O)O)([H])O[C@@H]7CO)C5(C)C)C)[C@]3(C)CC1)O[C@@H]([C@@H]([C@@H](O)[C@@H]8O)O)O[C@@H]8CO |
| Molecular Weight (g/mol) | 957.11 |
| Show More |