missing translation for 'onlineSavingsMsg'
Learn More

Ginkgolide B

Product Code. p-7041897
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15473379 10 mg 10mg
15483379 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473379 Supplier Thermo Scientific Alfa Aesar Supplier No. J60646.MA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Platelet-Activating Factor (PAF) antagonist | CAS: 15291-77-7 | C20H24O10 | 424.402 g/mol

Ginkgolide B is used as a potent PAF-R antagonist. Ginkgolide B speeds up PAF hydrolysis by promoting PAF-AH I α2 homodimer activity. Additionally, this compound has been reported to inhibit neutrophil degranulation in vitro. Inhibits superoxide production in vitro and inhibits bronchoconstriction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ginkgolide B is used as a potent PAF-R antagonist. Ginkgolide B speeds up PAF hydrolysis by promoting PAF-AH I α2 homodimer activity. Additionally, this compound has been reported to inhibit neutrophil degranulation in vitro. Inhibits superoxide production in vitro and inhibits bronchoconstriction.

Solubility
Soluble in DMSO. Slightly soluble in water and ethanol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15291-77-7
Molecular Formula C20H24O10
Molecular Weight (g/mol) 424.402
MDL Number MFCD05662347
InChI Key SQOJOAFXDQDRGF-WVXYHUDCSA-N
Synonym sqojoafxdqdrgf-maggbrlhsa-n
PubChem CID 132470872
SMILES CC1C(=O)OC2C1(C34C(=O)OC5C3(C2O)C6(C(C5)C(C)(C)C)C(C(=O)OC6O4)O)O

Specifications

Color White
Quantity 10 mg
Merck Index 14,4426
Solubility Information Soluble in DMSO; Slightly soluble in water and ethanol
Formula Weight 424.4
Physical Form Powder
Chemical Name or Material Ginkgolide B

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.