missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GIN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571571
This item is not returnable.
View return policy
Description
GIN1 Polyclonal Antibody for Western Blot, IHC (P)
GIN1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding; DNA integration.
Specifications
| GIN1 | |
| Polyclonal | |
| Unconjugated | |
| GIN1 | |
| 4930429M06Rik; 8430411H10Rik; Gin1; Gin-1; gypsy retrotransposon integrase 1; gypsy retrotransposon integrase-like protein 1; TGIN1; Ty3/Gypsy integrase 1; ZH2C2; Zinc finger H2C2 domain-containing protein; zinc finger, H2C2 domain containing | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 252876, 54826 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8K259, Q9NXP7 | |
| GIN1 | |
| KLH conjugated synthetic peptide between 464-492 amino acids from the C-terminal region of human GIN1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction