missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Neurobasal™ Plus Medium
GREENER_CHOICE

Product Code. 15886451
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15886451 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15886451 Supplier Gibco™ Supplier No. A3582901

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Neurobasal Plus Medium is a neuronal basal medium that provides the highest rates of in vitro survival of primary rodent and human stem cell-derived neurons when used with B-27 Plus Supplement. Neurobasal Plus Medium features an optimized formulation, upgraded manufacturing process, and more stringent quality control of raw materials and final product. These improvements enable the highest rates of neuronal cell survival, improved electrophysiological activity, and enhanced functional maturity versus other neuronal cell culture media systems.

Neurobasal Plus Medium, when combined with B-27 Plus Supplement, offers:

  • At least 50% higher survival rate of primary rat neurons compared to classic Neurobasal Medium and B-27 Supplement
  • Highest survival rate of primary rat, mouse, and human stem cell-derived neurons compared to other commercially available serum-free neuronal media systems
  • Improved electrophysiological activity and maturation—more consistent, stable, and synchronized spontaneous activity in primary rodent neurons after 10 days compared to BrainPhys™ Neuronal Medium
  • Accelerates neurite outgrowth and increases the number of synaptic-positive puncta

Available as a 500 mL basal medium, Neurobasal Plus Medium can seamLessly replace other neuronal cell culture media, such as classic Neurobasal Medium. The B-27 Plus Neuronal Culture System is also available for purchase, which is comprised of B-27 Plus Supplement and Neurobasal Plus Medium, offering a convenient and valuable all-in-one ordering solution.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Neural Cells
Classification Serum-free
Form Liquid
Product Type Neurobasal Medium
With Additives Phenol Red
Product Line Neurobasal
Quantity 500 mL
Shipping Condition Ambient
Tested For Endotoxin, Performance, Sterility, Osmolality, pH
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Neuronal Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.