missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carl Roth™ GHS Hazard Label
Click to view available options
Displayed Text:
Corrosion Pictogram/Achtung/Warning/Attention
Skull and Crossbones Pictogram/Gefahr/Danger/Danger
Unit Size:
Pack of 250
Description
- High-glossy PE-foil
- 100 μm thick
- Permanent adhesive
- Waterproof
- Temperature stable from -20 to +80 °C
- Signal words trilingual: German, English, French
- 250 per roll in practical dispenser box
Specifications
Specifications
| Certifications/Compliance | UN Regulation |
| Finish | Gloss |
| Material | Polyethylene |
| Quantity | 250/Pk. |
| Adhesive Type | Permanent |
| Height (Metric) | 30 mm |
| Thickness | 100 μ |
| Width (Metric) | 22 mm |
| Temperature Range (Metric) | -20°C to 80°C |
| Languages | French |
| Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction