Learn More
Ghrelin Receptor/GHSR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16655
This item is not returnable.
View return policy
Description
Ghrelin Receptor/GHSR Polyclonal specifically detects Ghrelin Receptor/GHSR in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GHSR | |
| 41 kDa | |
| RUO |
| Rabbit | |
| Affinity Purified |