missing translation for 'onlineSavingsMsg'
Learn More

GGTI-2418, MedChemExpress

Product Code. 16439557
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16439557

Brand: Medchem Express HY16231

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GGTI-2418 is a highly potent, competitive, and selective geranylgeranyltransferase I (GGTase I) inhibitor. GGTI-2418 inhibits GGTase I and FTase activities with IC50s of 9.5 nM and 53 μM, respectively. GGTI-2418 also increases p27(Kip1) and induces significant regression of breast tumors.

GGTI-2418 is a highly potent, competitive, and selective geranylgeranyltransferase I (GGTase I) inhibitor. GGTI-2418 inhibits GGTase I and FTase activities with IC50s of 9.5 nM and 53 μM, respectively. GGTI-2418 also increases p27(Kip1) and induces significant regression of breast tumors.

Specifications

Chemical Name or Material GGTI-2418
Physical Form Solid
CAS 501010-06-6
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C23H31N5O4
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 125 mg/mL (283.11 mM; Need ultrasonic)
SMILES CC(C)C[C@@H](C(O)=O)NC(N1[C@@H](CC2=CC=CC=C2)C(N(CC3=C(C)N=CN3)CC1)=O)=O
Molecular Weight (g/mol) 441.52
Formula Weight 441.52
Percent Purity 98.04%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.